Various Replacement Parts, Sciencix
Filter, Air, Acquity Pump, ACQUITY I-Class BSM, ACQUITY UPC2 BSM, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM, WATE, 700002632
Catalog # 103011-586
Marketsource
Supplier: SCIENCIX
Supplier #: CTS-11116
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Filter, Air, Acquity Pump
- OEM manufacturer:WATE
- OEM equivalent:700002632
- Model:ACQUITY I-Class BSM, ACQUITY UPC2 BSM, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM