Order Entry
United States
Orders LinkContactUsLinkComponent
Methoxyfenozide ≥98%

Methoxyfenozide ≥98%

Vial, 100 mg
Catalog # 102990-530
Environment
Marketsource
Supplier:   Adipogen
Supplier #:  CDX-M0133-M100
Methoxyfenozide ≥98%
Catalog # 102990-530
Supplier:  Adipogen
Marketsource
CAS Number:  161050-58-4
Methoxyfenozide ≥98%

Vial, 100 mg

CAS Number:  161050-58-4
Price
$91.38
Log in for availability
Each (100mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Diacylhydrazine insecticide. Ecdysone agonist that mimics the biological function of the natural insect molting hormone 20-hydroxyecdysone, causing premature molting and interfering with the normal growth and development in insects. Ecdysteroid agonist are also used as ligands for gene expression and gene therapy research. Compound can be used as analytical reference material.

Synonyms:  Methoxyphenozide
  • CAS Number:161050-58-4
  • Formula:C₂₂H₂₈N₂O₃
  • MDL Number:MFCD03792750
  • Molecular Weight:368.5 g/mol
  • Storage Temperature:Freezer
  • UN:3077
  • ADR:9,III
Specifications

Specification Test Results

  • Source/Host:
    Synthetic.
  • Purity:
    98% (HPLC)
  • Appearance/Color:
    White powder.
  • Solubility:
    Soluble in chloroform.

Specifications

  • Pack type:
    Vial
  • Size:
    100 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.