Order Entry
United States
Orders LinkContactUsLinkComponent
Anthralin

Anthralin

Vial, 1 g
Catalog # 102989-742
Marketsource
Supplier:   Adipogen
Supplier #:  CDX-D0308-G001
Anthralin
Catalog # 102989-742
Supplier:  Adipogen
Marketsource
CAS Number:  1143-38-0
Anthralin

Vial, 1 g

CAS Number:  1143-38-0
Price
$109.38
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Anthralin is a leukotriene biosynthesis inhibitor. It inhibits LTB4 omega-oxidation and disrupts mitochondria function. Anthralin is used in the treatment of psoriasis, as a fungicide, in the treatment of ringworm infections and in chronic dermatoses. It accumulates in mitochondria where it interferes with the supply of energy to the cell, probably by the oxidation of dithranol releasing free radicals. This impedes DNA replication and so slows the excessive cell division that occurs in psoriatic plaques. Numerous studies have demonstrated anti-proliferative and anti-inflammatory effects of anthralin on psoriatic and normal skin. It is also used as matrix substance for MALDI-MS (matrix-assisted laser desorption ionization MS).

Synonyms:  1,8,9-Trihydroxyanthracene, 1,8,9-Anthracenetriol, DIT, Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-one
  • CAS Number:1143-38-0
  • Formula:C₁₄H₁₀O₃
  • MDL Number:MFCD00053409
  • Molecular Weight:226.23 g/mol
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.