Orders LinkContactUsLinkComponent
Basic IV Administration Sets, B. Braun
Basic IV Administration Sets, B. Braun
Universal Spike, Roller Clamp, Slide Clamp, Injection Sites 26" Above Distal End, SPIN-LOCK® connector, Priming Volume: 19 ml, 15 Drops/ml, 236.2 cm (93"), Non sterile
Catalog # 10821-466
Supplier:   B Braun
Supplier #:  V1415-15
Basic IV Administration Sets, B. Braun
Catalog # 10821-466
Supplier:  B Braun
Basic IV Administration Sets, B. Braun
Basic IV Administration Sets, B. Braun
Universal Spike, Roller Clamp, Slide Clamp, Injection Sites 26" Above Distal End, SPIN-LOCK® connector, Priming Volume: 19 ml, 15 Drops/ml, 236.2 cm (93"), Non sterile
Price
$317.50
Log in for availability
Case of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
B. Braun gravity sets are available in various lengths and dimensions to accommodate various applications.
Specifications
  • Flow rate:
    15 Drops/ml
  • Sterility:
    Non sterile
  • Description:
    Universal Spike, Roller Clamp, Slide Clamp, Injection Sites 26" Above Distal End, SPIN-LOCK® connector, Priming Volume: 19 ml
  • Length:
    236.2 cm (93")