Order Entry
United States
Orders LinkContactUsLinkComponent
 

Human Recombinant CD80

50 μg
  102970-306
Marketsource
 :  CK61-50UG
Human Recombinant CD80
  102970-306
Marketsource
 :  
Human Recombinant CD80

50 μg

 :  
 
$184.54
   
Each (50µG)
 
   
 

Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products

  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels

Recombinant Human Activation B7-1 antigen is produced by our Mammalian expression system and the target gene encoding Val35-Asn242 is expressed with a 6His tag at the C-terminus.

 : Accessories not included.

 : Shipped Via FedEx

 : Shipped at ambient temperature via FedEx envelope.

 : This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.

The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.

 

 

  • Size:
    50 μg
  • Cat. No.:
    102970-306
  • Protein/Peptide Type:
    Recombinant
  • Protein/Peptide Name:
    CD80
  • Species:
    Human

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.