COMPLEMENT PROTEIN HUMAN C3 CONC 5MG/ML
Catalog # MSPP-A113C
Supplier: COMPTECH MS
COMPLEMENT PROTEIN HUMAN C3 CONC 5MG/ML
Catalog # MSPP-A113C
Supplier: COMPTECH MS
Supplier Number: A113C
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Multipurpose Containers, HDPE
Supplier: VWR International
Technical Buffer Solutions, Sachets
Supplier: Mettler Toledo
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
Reynolds Wrap® Aluminum Foil
Supplier: Control Company
Tissue-Tek® Cryomold® Molds/Adapters, Sakura® Finetek
Supplier: Sakura Finetek
SOLUTION BOND EPITOPE RETRIEVAL 1L
Supplier: Leica Biosystems MS
ASSAY RPLEX HUMAN COMPLEMENT K151V0R-2
Supplier: Meso Scale Diagnostics
High Sensitivity RNA ScreenTape Analysis
Supplier: Agilent Technologies
FACTOR P 1MG/ML 100UG
Supplier: COMPTECH MS
RediSep Rf Gold Silica Flash Chromatography Column, Teledyne ISCO
Supplier: Teledyne
BioProfile® Flex 2 Microsensor Cards for Automated Chemistry Analyzers, Nova Biomedical
Supplier: Nova Biomedical
HEPES Buffer, 1M, Teknova
Supplier: Teknova
BioBall®, MultiShot 550, bioMerieux
Supplier: bioMerieux
Nunc® 384-Well Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
TRANSFECT REAGENT I 10MG
Supplier: Avanti Polar Lipids Inc
MARKER 6.14 MAURICE CIEF PI 210UL
Supplier: 0000014126
FACTOR D 0.1MG/ML 25UG
Supplier: COMPTECH MS