Order Entry
United States
Orders LinkContactUsLinkComponent
Vernier® Salinity Sensor

Vernier® Salinity Sensor

Vernire salinitysStandard 35 ppt, 500 ml
Catalog # 470219-326
Supplier #:  SAL-ST
Vernier® Salinity Sensor
Catalog # 470219-326
Vernier® Salinity Sensor
Vernier® Salinity Sensor

Vernire salinitysStandard 35 ppt, 500 ml

Price
$26.29
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Easily and precisely measure the total dissolved salt content in ocean or brackish water.

Response time: 90% of full-scale reading in 10 s.

Specifications
  • Description:
    Vernire salinitysStandard 35 ppt, 500 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.