CELL LINE U-2 OS
Catalog # MSPP-HTB96
Supplier: 0000013421
Frequently Bought Together
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
Minncare® Test Strips, Mar Cor Purification
Supplier: Mar Cor Purification
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Corning® Centrifuge Tubes, 15 ml
Supplier: Corning
CiDehol® ST Sterile 70% IPA with WFI, Decon Labs
Supplier: Decon Labs
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Cryo.s™ Cryogenic Storage Vials, Polypropylene, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
PROTEIN STANDARD II F/MASS SPECTROMETRY
Supplier: 0000019826
ATCC® CRL-3216™ 293T Cell Line, American Type Culture Collection
Supplier: 0000013421
Lok-Tight™ Contact Plates, Hardy Diagnostics
Supplier: Hardy Diagnostics
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar
Supplier: Hardy Diagnostics
STEM CELL ADIPOSEDERIVED MESENCHYMAL HMN
Supplier: 0000013421
PRIMARY UMBILICAL VEIN ENDOTHELIAL CELLS
Supplier: 0000013421