Basic IV Administration Sets, B. Braun
Product Details & Documents
B. Braun gravity sets are available in various lengths and dimensions to accommodate various applications.
Documents
Specifications
- Flow rate:20 Drops/ml
- Description:Universal Spike with 15 µm Filter, on/off Clamp, Rate Flow Regulator, Injection Site 6" Above Distal End, SPIN-LOCK® Connector, Priming Volume: 16.5 ml
- Length:211.5 cm (83.27")