Order Entry
United States
Orders LinkContactUsLinkComponent
 

Envelope Vectors, Cell Biolabs

pCMV-Ampho, 100 µL
Catalog # 102965-762
Marketsource
Envelope Vectors, Cell Biolabs
Catalog # 102965-762
Supplier:  
Marketsource
CAS Number:  
Envelope Vectors, Cell Biolabs
Envelope Vectors, Cell Biolabs

pCMV-Ampho, 100 µL

CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cell Biolabs' Envelope Vectors each contain a different envelope gene for pseudotyping recombinant lentivirus or retrovirus

Co-transfect one of these constructs along with other packaging and cloning vectors into 293T or 293RTV cells.

Specifications

Specifications

  • Description:
    pCMV-Ampho
  • Size:
    100 µL
  • Cat. No.:
    102965-762
  • Format:
    Bacterial Glycerol Stock
  • Storage Temperature:
    –80°C

Specifications