Order Entry
United States
Orders LinkContactUsLinkComponent
AAV-5 Helper Free Expression Complete Systems, Cell Biolabs

AAV-5 Helper Free Expression Complete Systems, Cell Biolabs

AAV-5 Bicistronic Expression System (Blasticidin), 1 Kit
Catalog # 102965-552
Marketsource
Supplier #:  VPK-419-SER5
AAV-5 Helper Free Expression Complete Systems, Cell Biolabs
Catalog # 102965-552
Supplier:  
Marketsource
CAS Number:  
AAV-5 Helper Free Expression Complete Systems, Cell Biolabs
AAV-5 Helper Free Expression Complete Systems, Cell Biolabs

AAV-5 Bicistronic Expression System (Blasticidin), 1 Kit

CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

AAV-5 Helper Free Expression Complete Systems contain the following constructs needed to package your recombinant serotype-5 adeno-associated virus: an expression vector, pHelper plasmid, a serotype-specific Rep-Cap plasmid, and an optional GFP expression vector for making a control virus

AAV Helper Free Expression Complete Systems are available for native serotypes 1 through 6. The shRNA expression vectors contain a U6 promoter. All other expression vectors contain a CMV promoter which is optimal for general gene expression, with the exception of our promoterless vector which is useful if you wish to clone in a gene-specific promoter along with your gene of interest.

Specifications

Specifications

  • Description:
    AAV-5 Bicistronic Expression System (Blasticidin)
  • Size:
    1 Kit
  • Cat. No.:
    102965-552
  • Format:
    Purified Plasmid
  • Storage Temperature:
    –20°C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.