RIPA Buffer, Cell Biolabs
RIPA Buffer, 20mL
Catalog # 102963-646
Marketsource
Supplier: Cell Biolabs
Supplier #: AKR-191
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Cell Biolabs' RIPA Buffer is suitable for the extraction of cytoplasmic, membrane, and nuclear proteins from cells or tissues
The buffer is suitable for a variety of downstream applications such as reporter assays, protein assays, immunoassays and protein purification. RIPA Buffer is available with or without a separate Protease Inhibitor Cocktail.
Specifications
Specifications
- Description:RIPA Buffer
- Size:20mL
- Cat. No.:102963-646
- Storage Temperature:Room Temperature and –20°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



