ContactUsLinkComponent
Microorganism Tests, LPS from Escherichia coli O111:B4
Lipopolysaccharides (LPS) and Related Products, List Biological Laboratories
Catalog # 102946-484
CAS Number:  
Lipopolysaccharides (LPS) and Related Products, List Biological Laboratories
Catalog # 102946-484
Supplier Number:  201
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    LPS from Escherichia coli O111:B4
  • Size:
    5 mg
  • Environmentally Preferable:
  • Cat. No.:
    102946-484
  • Supplier no.:
    201
Specifications
About this item
Bacterial lipopolysaccharides have long been recognized as the active component of gram negative bacterial endotoxins
LPS from Escherichia coli O111:B4 is a smooth strain of LPS that is isolated by a modification of the phenol extraction method of Westphal and Jann. Smooth strain LPS are dispersable in aqueous solvents at concentrations of up to 5.0 mg/ml.
These unique macromolecules have been extensively studied by investigators in many disciplines in efforts to elucidate and define relevant pathophysiologival parameters of endotoxin shock, a profound life-threatening consequence of bacterial sepsis. Lipopolysaccharides have generated intense interest as being among the most potent natural products capable of pluripotential immunostimulation manifested by the activation of host cells (e.g. B. lymphocytes, macrophages) to functional differentiation. Host cell activation by lipopolysaccharides produces a spectrum of hormone-active lymphokines and monokines, including interferons (alpha, beta and gamma), interleukins 1 and 6, tumor necrosis factor, platelet activating factor and procoagulant tissue factor. The documented capacity of lipopolysaccharids or their active lipid A component to initiate a variety of biochemical pathways (protein kinase C, cAMP dependent protein kinase, phosphatidyl inositol turnover, arachidonate metabolism, protein myristolation and activation of G-proteins) provides investigators with powerful molecular tools by which to study cellular activation mechanisms.
Frequently Bought Together