Order Entry
United States
Orders LinkContactUsLinkComponent
 

Methanol ≥99%

500 ml
Catalog # BT223840-500ML
Supplier:   BeanTown Chemical
 :  223840-500ML
Methanol ≥99%
Catalog # BT223840-500ML
Supplier:  BeanTown Chemical
CAS Number:  67-56-1
Methanol ≥99%
Methanol ≥99%

500 ml

CAS Number:  67-56-1
Price
$23.90
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 67-56-1; EC No: 200-659-6; MDL No: MFCD00004595; RTECS: PC1400000 UN No: UN1230; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3OH; MW: 32.04 Melting Point: -98°; Boiling Point: 64.7°; Flash point: 9.7°C (49.5°F) Density (g/mL): 0.791; Refractive Index: 1.329

  • RTECS: PC1400000
  • Refractive index: 1.329
  • Liquid
Synonyms:  Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Boiling Pt:64.7 °C
  • Melting Pt:–98 °C
  • Density:0.791 g/mL
  • Flash Pt:9.7 °C (49.5 °F)
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
Specifications

Specification Test Results

  • Assay:
    99%

Specifications

  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.