Order Entry
United States
Orders LinkContactUsLinkComponent
Lithium hydroxide monohydrate, Technical Grade

Lithium hydroxide monohydrate, Technical Grade

2 kg
Catalog # BT219170-2KG
Supplier:   BeanTown Chemical
Supplier #:  219170-2KG
Lithium hydroxide monohydrate, Technical Grade
Catalog # BT219170-2KG
Supplier:  BeanTown Chemical
CAS Number:  1310-66-3
Lithium hydroxide monohydrate, Technical Grade

2 kg

CAS Number:  1310-66-3
Price
$695.00
Log in for availability
Each (2kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 1310-66-3; EC No: 215-183-4; MDL No: MFCD00149772; RTECS: OJ6307080 UN No: UN2680; Haz Class: 8; Packing Group: II Powder; Linear Formula: LiOH·H2O ; MW: 41.96 Density (g/mL): 1.51 Air Sensitive , Hygroscopic

  • RTECS: OJ6307080
  • Air Sensitive, Hygroscopic
  • Powder
  • CAS Number:1310-66-3
  • Formula:LiOH·1H₂O
  • MDL Number:MFCD00149772
  • Molecular Weight:41.96 g/mol
  • Density:1.51 g/mL
  • EINECS:215-183-4
  • UN:2680
  • ADR:8,II

Documents

Specifications

Specifications

  • Size:
    2 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.