Order Entry
United States
Orders LinkContactUsLinkComponent
Acetone ≥99.5% for HPLC

Acetone ≥99.5% for HPLC

1 L
Catalog # BT128560-1L
   Safer Human and Environmental Health
Supplier:   BeanTown Chemical
Supplier #:  128560-1L
Acetone ≥99.5% for HPLC
Catalog # BT128560-1L
Supplier:  BeanTown Chemical
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone ≥99.5% for HPLC
Acetone ≥99.5% for HPLC

1 L

CAS Number:  67-64-1
Price
$48.30
Log in for availability
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 67-64-1; EC No: 200-662-2; MDL No: MFCD00008765; RTECS: AL3150000 UN No: UN1090; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3COCH3; MW: 58.08 Melting Point: -94°; Boiling Point: 56°; Flash point: -17°C (1°F) Density (g/mL): 0.791; Refractive Index: 1.359

  • RTECS: AL3150000
  • Refractive index: 1.359
  • Liquid
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Boiling Pt:56 °C
  • Melting Pt:–94 °C
  • Density:0.791 g/mL
  • Flash Pt:–17 °C (1 °F)
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Assay:
    99.5% min.

Specifications

  • Size:
    1 L
  • Cat. No.:
    BT128560-1L
  • Environmental Certification:
    Chem21 Solvent selection guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.