Order Entry
United States
Orders LinkContactUsLinkComponent
Sodium chlorite ≥80%, Technical Grade

Sodium chlorite ≥80%, Technical Grade

2 kg
Catalog # BT127350-2KG
Supplier:   BeanTown Chemical
Supplier #:  127350-2KG
Sodium chlorite ≥80%, Technical Grade
Catalog # BT127350-2KG
Supplier:  BeanTown Chemical
CAS Number:  7758-19-2
Sodium chlorite ≥80%, Technical Grade

2 kg

CAS Number:  7758-19-2
Price
$98.00
Log in for availability
Each (2kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 7758-19-2; EC No: 231-836-6; MDL No: MFCD00003478; RTECS: VZ4800000 UN No: UN1496; Haz Class: 5.1; Packing Group: II Powder; Linear Formula: NaClO2; MW: 90.44 Melting Point: 180-200° (decomposes) Hygroscopic

  • RTECS: VZ4800000
  • Hygroscopic
  • Powder
  • CAS Number:7758-19-2
  • Formula:ClNaO₂
  • MDL Number:MFCD00003478
  • Molecular Weight:90.44 g/mol
  • Melting Pt:180…200 °C (dec.)
  • EINECS:231-836-6
  • UN:1496
  • ADR:5.1,II

Documents

Specifications

Specification Test Results

  • Assay:
    80%

Specifications

  • Size:
    2 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.