Orders LinkContactUsLinkComponent
MULTIVIEW® Universal Tissue Microarray, Enzo Life Sciences
MULTIVIEW® Universal Tissue Microarray, Enzo Life Sciences
25 slides
Catalog # 10014-538
Supplier:   Enzo Life Sciences
Supplier #:  950-020-0025
MULTIVIEW® Universal Tissue Microarray, Enzo Life Sciences
Catalog # 10014-538
Supplier:  Enzo Life Sciences
MULTIVIEW® Universal Tissue Microarray, Enzo Life Sciences
25 slides
Price
$893.81
Log in for availability
Each
Product Details & Documents
MULTIVIEW® Universal tissue microarray with 11 different human tissue specimens for a broad-spectrum of immunohistochemical staining procedures.
Specifications
  • Size:
    25 slides