Orders LinkContactUsLinkComponent
Protector® Pass-Through Laboratory Hoods, Labconco®
Protector® Pass-Through Laboratory Hoods, Labconco®
Hood with Right-Side Window, 230V, 33.4", 59", 0, 2, 775 cfm, 590 cfm, 420 lbs.
Catalog # 10799-248
CSA Certified
Product available on GSA Advantage®
UL Listed
   Sustainable Materials
Supplier:   Labconco
Supplier #:  113408021
Protector® Pass-Through Laboratory Hoods, Labconco®
Catalog # 10799-248
Supplier:  Labconco
CSA Certified
Product available on GSA Advantage®
UL Listed
   Sustainable Materials
Protector® Pass-Through Laboratory Hoods, Labconco®
Protector® Pass-Through Laboratory Hoods, Labconco®
Hood with Right-Side Window, 230V, 33.4", 59", 0, 2, 775 cfm, 590 cfm, 420 lbs.
Price
$12,720.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Laboratory hoods feature sashes on two sides that, when mounted in an island or peninsular configuration, provide unobstructed viewing from either side during chemistry demonstrations or observing students using the hood
Ergonomic air foils allow air to sweep the work surface for maximum containment. Clean-Sweep* airflow openings pull inflow air from under the airfoil so that clean air continually flows over the airfoil, creating a constant barrier of protection from contaminants. Upper baffle efficiently directs airflow to the interior center. Bypass airflow design ensures relatively stable face velocities. The 25.6cm (103/32") I.D. exhaust connection is made of powder-coated steel.
Durable frame is constructed of powder-coated steel and aluminum. Chemical-resistant, fiberglass-reinforced composite panel liner surpasses all national codes for flame spread and has a bright white surface for excellent light reflectivity. Front and side panels may be easily removed for lamp replacement and access to sash weight and electrical supply connections. Clear, vertical-rising tempered safety glass sashes and streamlined corner posts provide maximum visibility. An optional 38.1Wx61Hcm (15x24") tempered, safety glass viewing window can be mounted on the left or right side for enhanced visibility. An incandescent light, switches, and a GFCI electrical duplex receptacle are factory pre-wired to the hood's junction box. Color: glacier white.
SpillStopper Work Surfaces (sold separately) are solid, black surfaces molded of a special formulation of corrosion-resistant epoxy resin. They are dished and contoured to conform to interior liner of Protector Pass-Through Hoods. Pre-drilled 2.5" (6.4cm) dia plumbing holes. May be ordered with pre-cut 6" x 3" (15.2 x 7.6 cm) oval cupsink cutout(s). Cupsink is sold separately.
These hoods may also be installed in the wall between two rooms. An audible alarm sounds when both sashes are opened at the same time.
 : Hoods meet SEFA 1, ASHRAE 110-95, ANSI Z9.5, UL 3101-1/61010-1, and CAN/CSA C22.2 No. 1010.1 standards. ETL listed.
 : Hoods require use of a remote blower, ductwork, work surface, and base cabinet (all sold separately); contact your Avantor sales representative for more information.
 : VWR CATALYST™ offers customer on-site validation, Testing & certification services including Face Velocity Profile Test, Smoke Generation Test, Tracer Gas Test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWR[email protected] for more details. You can also visit https://vwr.com/equip_services.
Specifications
  • Description:
    Hood with Right-Side Window, 230V
  • No. of service fixtures:
    2
  • No. of duplex receptacles:
    0
  • Total exhaust volume at 100 fpm (Sash 100% open):
    775 cfm
  • Total exhaust volume at 100 fpm (Sash 60% open):
    590 cfm
  • Outside height:
    59"
  • Outside depth:
    33.4"
  • Shipping weight:
    420 lbs.
  • Cat. No.:
    10799-248
  • Environmentally Preferable:
    Sustainable Materials
Accessories
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Right Rear Cupsink Cutout
    SpillStopper Work Surface, Right Rear Cupsink Cutout
    10805-110
    4' Pass-Through Hoods
    Log in for availability
    Each - $2,120.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left Side Cupsink Cutout
    SpillStopper Work Surface, Left Side Cupsink Cutout
    10805-120
    4' Pass-Through Hoods
    Log in for availability
    Each - $2,120.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Right Side Cupsink Cutout
    SpillStopper Work Surface, Right Side Cupsink Cutout
    10805-112
    4' Pass-Through Hoods
    Log in for availability
    Each - $2,120.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Right Rear and Left Side Cupsink Cutouts
    SpillStopper Work Surface, Right Rear and Left Side Cupsink Cutouts
    10805-122
    4' Pass-Through Hoods
    Log in for availability
    Each - $2,350.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface
    SpillStopper Work Surface
    10805-108
    4' Pass-Through Hoods
    Log in for availability
    Each - $1,890.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left Rear Cupsink Cutout
    SpillStopper Work Surface, Left Rear Cupsink Cutout
    10805-114
    4' Pass-Through Hoods
    Log in for availability
    Each - $2,120.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left and Right Rear Cupsink Cutouts
    SpillStopper Work Surface, Left and Right Rear Cupsink Cutouts
    10805-116
    4' Pass-Through Hoods
    Log in for availability
    Each - $2,350.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left and Right Side Cupsink Cutouts
    SpillStopper Work Surface, Left and Right Side Cupsink Cutouts
    10805-124
    4' Pass-Through Hoods
    Log in for availability
    Each - $2,350.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left Rear and Right Side Cupsink Cutouts
    SpillStopper Work Surface, Left Rear and Right Side Cupsink Cutouts
    10805-118
    4' Pass-Through Hoods
    Log in for availability
    Each - $2,350.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface
    SpillStopper Work Surface
    10805-126
    5' Pass-Through Hoods
    Log in for availability
    Each - $2,370.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Right Rear Cupsink Cutout
    SpillStopper Work Surface, Right Rear Cupsink Cutout
    10805-128
    5' Pass-Through Hoods
    Log in for availability
    Each - $2,590.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Right Side Cupsink Cutout
    SpillStopper Work Surface, Right Side Cupsink Cutout
    10805-146
    5' Pass-Through Hoods
    Log in for availability
    Each - $2,590.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left and Right Rear Cupsink Cutouts
    SpillStopper Work Surface, Left and Right Rear Cupsink Cutouts
    10805-152
    5' Pass-Through Hoods
    Log in for availability
    Each - $2,820.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left Rear Cupsink Cutout
    SpillStopper Work Surface, Left Rear Cupsink Cutout
    10805-150
    5' Pass-Through Hoods
    Log in for availability
    Each - $2,590.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left Rear and Right Side Cupsink Cutouts
    SpillStopper Work Surface, Left Rear and Right Side Cupsink Cutouts
    10805-162
    5' Pass-Through Hoods
    Log in for availability
    Each - $2,820.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left Side Cupsink Cutout
    SpillStopper Work Surface, Left Side Cupsink Cutout
    10805-164
    5' Pass-Through Hoods
    Log in for availability
    Each - $2,590.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Right Rear and Left Side Cupsink Cutouts
    SpillStopper Work Surface, Right Rear and Left Side Cupsink Cutouts
    10805-166
    5' Pass-Through Hoods
    Log in for availability
    Each - $2,820.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left and Right Side Cupsink Cutouts
    SpillStopper Work Surface, Left and Right Side Cupsink Cutouts
    10805-168
    5' Pass-Through Hoods
    Log in for availability
    Each - $2,820.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface
    SpillStopper Work Surface
    10805-170
    6' Pass-Through Hoods
    Log in for availability
    Each - $2,820.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Right Rear Cupsink Cutout
    SpillStopper Work Surface, Right Rear Cupsink Cutout
    10805-172
    6' Pass-Through Hoods
    Log in for availability
    Each - $3,010.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Right Side Cupsink Cutout
    SpillStopper Work Surface, Right Side Cupsink Cutout
    10805-226
    6' Pass-Through Hoods
    Log in for availability
    Each - $3,010.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left Rear Cupsink Cutout
    SpillStopper Work Surface, Left Rear Cupsink Cutout
    10805-228
    6' Pass-Through Hoods
    Log in for availability
    Each - $3,010.00
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • SpillStopper Work Surface, Left and Right Rear Cupsink Cutouts
    SpillStopper Work Surface, Left and Right Rear Cupsink Cutouts
    10805-230
    6' Pass-Through Hoods
    Log in for availability
    Each - $3,230.00