SDS Binders
SDS Binder with 36" Metal Chain, 50 Protectors/Tabs, Holds up to 600 pages, 3"
Catalog # 470165-246
Supplier: EMEDCO
Supplier #: SDSB9
Jump to:
Product Details & Documents
SDS Binders keep essential SDS information organized and safe.
- Binders are clearly labeled
- Sheet protectors available
- SDS info safeguarded
The clearly labeled binders make it easy to store and access information.
They are available with and without sheet protectors. It is a sturdy plastic binder protects sheets from use and abuse.
Specifications
- Sheet capacity:Holds up to 600 pages
- Description:SDS Binder with 36" Metal Chain, 50 Protectors/Tabs
- Size:3"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



