Order Entry
United States
Orders LinkContactUsLinkComponent
Rose Bengal

Rose Bengal

25 g
Catalog # BT221180-25G
Supplier:   BeanTown Chemical
Supplier #:  221180-25G
Rose Bengal
Catalog # BT221180-25G
Supplier:  BeanTown Chemical
CAS Number:  632-69-9
Rose Bengal

25 g

CAS Number:  632-69-9
Price
$53.60
Log in for availability
Each (25g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 632-69-9; EC No: 211-183-3; MDL No: MFCD00151169; RTECS: LM5920000 Solid; Molecular Formula: C20H2Cl4I4Na2O5; MW: 1017.65 Melting Point: 300° Light Sensitive

  • RTECS: LM5920000
  • Light Sensitive
  • Solid
Synonyms:  Sodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Food Red No. 105, CI 45440, Acid red 94, Aniline Rose, Rose Bengal sodium
  • CAS Number:632-69-9
  • Formula:C₂₀H₂Cl₄I₄O₅Na₂
  • MDL Number:MFCD00151169
  • Molecular Weight:1017.63 g/mol
  • Melting Pt:>300 °C
  • EINECS:211-183-3

Documents

Specifications

Specifications

  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.