Order Entry
United States
Orders LinkContactUsLinkComponent
Lanolin

Lanolin

250 g
Catalog # BT144255-250G
Supplier:   BeanTown Chemical
Supplier #:  144255-250G
Lanolin
Catalog # BT144255-250G
Supplier:  BeanTown Chemical
CAS Number:  8006-54-0
Lanolin

250 g

CAS Number:  8006-54-0
Price
$43.30
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 8006-54-0; EC No: 232-348-6; MDL No: MFCD00081740; RTECS: OE3201000 Paste

  • RTECS: OE3201000
  • Paste
Synonyms:  Wool fat
  • CAS Number:8006-54-0
  • MDL Number:MFCD00081740
  • EINECS:232-348-6

Documents

Specifications

Specifications

  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.