Orders LinkContactUsLinkComponent
Oxoid CampyGen Compact
Oxoid CampyGen Compact
CampyGen compact, 20 sachets
Catalog # 10790-378
Product available on GSA Advantage®
Supplier:   Hardy Diagnostics
Supplier #:  CN020C
Oxoid CampyGen Compact
Catalog # 10790-378
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
Oxoid CampyGen Compact
Oxoid CampyGen Compact
CampyGen compact, 20 sachets
Price
$128.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Simple-to-use systems for the rapid generation of a microaerophilic environment necessary to cultivate Campylobacter species.
The Oxoid CampyGen and CampyGen Compact are simple-to-use systems for the rapid generation of a microaerophilic environment necessary to cultivate Campylobacter species.
Like the AnaeroGen, the CampyGen also requires no catalyst and no water activation steps. No dangerous gas is produced. The final atmosphere is reduced to 6% oxygen, and supplemented with 14% carbon dioxide which is ideal for growth of Campylobacter and other microaerophilic organisms. Sachet, bags and clips are sold separately.
Ordering information: Pouches and clips sold separately.
Specifications
  • Description:
    CampyGen compact
  • Size:
    20 sachets
  • Cat. No.:
    10790-378