Human FGF-basic Standard ABTS ELISA Development Kit, PeproTech, Inc.
Human FGF-basic ELISA Kit, Kit components sufficient to assay 10 plates
Catalog # 10777-634
Supplier: Peprotech
Supplier #: 900-K08K
Human FGF-basic Standard ABTS ELISA Development Kit, PeproTech, Inc.
Catalog # 10777-634
Supplier: Peprotech
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
PeproTech's Human FGF-basic ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human FGF-basic in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human FGF-basic ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human FGF-basic in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human FGF-basic approximately 1000 ELISA plate wells.
Specifications
Specifications
- Description:Human FGF-basic ELISA Kit
- Size:Kit components sufficient to assay 10 plates
- Cat. No.:10777-634
- Primary antibody reactivity:Human