Anti-RELMbeta Rabbit Polyclonal Antibody (Biotin)
Catalog # 10776-972
Supplier: Peprotech
Specifications
- Antibody Type:Secondary
- Antigen Name:RELMbeta
- Antigen Symbol:RELMbeta
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:25 µg
- Form:Lyophilized
- Storage Buffer:See Product Data Sheet for more information.
- Molecular Weight:19.0 kDa
- Storage Temperature:Store at –20 °C prior to reconstitution.
- Shipping Temperature:Ambient
- Immunogen:Recombinant Human RELMβ
- Purification:Recombinant Human RELMβ
- Cat. No.:10776-972
- Packaging:PP Vial
- Supplier no.:500P217BT25UG
Specifications
About this item
Produced from sera of rabbits immunized with highly pure Recombinant Human RELMβ. AntiHuman RELMβspecific antibody was purified by affinity chromatography and then biotinylated.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Secondary
Antigen: RELMbeta
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Frequently Bought Together