Order Entry
United States
Orders LinkContactUsLinkComponent
Anti-FGF-basic Rabbit Polyclonal Antibody

Anti-FGF-basic Rabbit Polyclonal Antibody

1 mg, PP Vial
Catalog # 10775-086
Supplier:   Peprotech
Supplier #:  500-P18-1MG
Anti-FGF-basic Rabbit Polyclonal Antibody
Catalog # 10775-086
Supplier:  Peprotech
Anti-FGF-basic Rabbit Polyclonal Antibody

1 mg, PP Vial

Price
$3,028.31
Log in for availability
Each (1mg)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Produced from sera of rabbits immunized with highly pure Recombinant Human FGF-basic. Anti­Human FGF-basic­specific antibody was purified by affinity chromatography employing an immobilized Human FGF-basic matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: FGF-basic
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Specifications
  • Size:
    1 mg
  • Packaging:
    PP Vial
  • Clonality:
    Polyclonal
  • Molecular Weight:
    17.2 kDa
  • Purification:
    Recombinant Human FGF-basic
  • Form:
    Lyophilized
  • Storage Buffer:
    See Product Data Sheet for more information.
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Cat. No.:
    10775-086
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Shipping Temperature:
    Ambient
  • Antibody Type:
    Primary
  • Antigen Symbol:
    FGF-basic
  • Immunogen:
    Recombinant Human FGF-basic
  • Conjugation:
    Unconjugated
  • Antigen Name:
    FGF-basic
  • ELISA:
    Yes
  • Storage Temperature:
    Store at –20 °C prior to reconstitution.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.