Anti-FGF-basic Rabbit Polyclonal Antibody
50 µg, PP Vial
Catalog # 10775-082
Supplier: Peprotech
Supplier #: 500-P18-50UG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Produced from sera of rabbits immunized with highly pure Recombinant Human FGF-basic. AntiHuman FGF-basicspecific antibody was purified by affinity chromatography employing an immobilized Human FGF-basic matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: FGF-basic
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Size:50 µg
- Packaging:PP Vial
- Clonality:Polyclonal
- Molecular Weight:17.2 kDa
- Purification:Recombinant Human FGF-basic
- Form:Lyophilized
- Storage Buffer:See Product Data Sheet for more information.
- Isotype:IgG
- Host:Rabbit
- Cat. No.:10775-082
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:Ambient
- Antibody Type:Primary
- Antigen Symbol:FGF-basic
- Immunogen:Recombinant Human FGF-basic
- Conjugation:Unconjugated
- Antigen Name:FGF-basic
- ELISA:Yes
- Storage Temperature:Store at –20 °C prior to reconstitution.



