Order Entry
United States
Orders LinkContactUsLinkComponent
Mouse Recombinant SCF (from <i>E. coli</i>)

Mouse Recombinant SCF (from E. coli)

10 µg, US
Catalog # 10771-728
Supplier:   Peprotech
Supplier #:  AF-250-03-10UG
Mouse Recombinant SCF (from <i>E. coli</i>)
Catalog # 10771-728
Supplier:  Peprotech
CAS Number:  
Mouse Recombinant SCF (from E. coli)

10 µg, US

CAS Number:  
Price
$455.47
Log in for availability
Each (10µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells. The human SCF gene encodes for a 273 amino acid transmembrane protein, which contains a 25 amino acid N-terminal signal sequence, a 189 amino acid extracellular domain, a 23 amino acid transmembrane domain, and a 36 amino acid cytoplasmic domain. The secreted soluble form of SCF is generated by proteolytic processing of the membrane anchored precursor. Recombinant Murine SCF is an 18.3 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF. Manufactured using all Animal-Free reagents.

  • Carrier free
  • Animal-free

MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA

Verified by N-terminal and Mass Spectrometry analyses (when applicable).

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Specifications

Specifications

  • Size:
    10 µg
  • Origin:
    US
  • Storage Conditions:
    −20 °C
  • Cat. No.:
    10771-728
  • Shipping Temperature:
    RT
  • Biological Activity:
    The ED<sub>50</sub> was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10<sup>5</sup> units/mg.
  • Protein/Peptide Type:
    Recombinant
  • Formulation:
    Lyophilized
  • Animal-Free:
    Y
  • Purity:
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Protein/Peptide Name:
    SCF
  • Endotoxin Content:
    <0.1 ng/μg of protein (<1 EU/μg).
  • Species:
    Mouse
  • Source:
    E. coli

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.