Order Entry
United States
Orders LinkContactUsLinkComponent
NEFA Linearity Set Solution, Wako

NEFA Linearity Set Solution, Wako

NEFA Linearity Set, 1×10 ml, +2…+10 °C
  10753-190
Supplier #:  997-76491
NEFA Linearity Set Solution, Wako
  10753-190
 :  
NEFA Linearity Set Solution, Wako
NEFA Linearity Set Solution, Wako

NEFA Linearity Set, 1×10 ml, +2…+10 °C

 :  
 
$185.84
   
Each (10ml)
 
   
 

The NEFA Linearity Set is a 2.5 mEq/L solution used to verify the measurable range of the NEFA HR(2) assay.

This product is only used with Wako’s NEFA HR(2) series of reagents.

Documents

 

 

  • Description:
    NEFA Linearity Set
  • Size:
    1×10 ml
  • Storage temperature:
    +2…+10 °C

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.