Sample Loop with Fingertight Connectors, Cytiva
Sample Loop, Polyetheretherketone (PEEK), 1.0 ml, 0.75 mm (0.029"), 1.58 mm (¹⁄₁₆") fitted with fingertight connectors
Catalog # 10497-998
Supplier: Cytiva
: 18-1114-01
Jump to:
Product Details & Documents
The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100% Ethyl acetate, 100% Hexane, and 100% Tetrahydrofuran.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Tubing ext.Ø:1.58 mm (¹⁄₁₆") fitted with fingertight connectors
- Tubing int.Ø:0.75 mm (0.029")
- Capacity:1.0 ml
- Description:Sample Loop
- Material:Polyetheretherketone (PEEK)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



