Order Entry
United States
Orders LinkContactUsLinkComponent
 

VWR® Buffer Solutions

Phosphate, Poly bottle, 4 L, 7.00, Green
Catalog # 71006-774
 :  ANAC15229-540
VWR® Buffer Solutions
Catalog # 71006-774
Supplier:  
CAS Number:  
VWR® Buffer Solutions
VWR® Buffer Solutions

Phosphate, Poly bottle, 4 L, 7.00, Green

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BDH BUFFER REF STD PH7 YELLOW 4L Catalog #BDH5048-4L

 

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These buffer solutions are for calibrating pH meters

They are traceable to NIST standards.

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Color:
    Green
  • Size:
    4 L
  • pH buffer type:
    Phosphate
  • pH:
    7.00

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.