Orders LinkContactUsLinkComponent
Micro Trays, Sklar®
Micro Trays, Sklar®
8×14×11/4"
Catalog # 10193-238
Micro Trays, Sklar®
Catalog # 10193-238
Supplier:  
Micro Trays, Sklar®
Micro Trays, Sklar®
8×14×11/4"
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Micro Sterilizing Trays are used to hold smaller instruments securely during a sterilization process. Rows of separating pins and silicone hold down bars for locking instruments in place. The tray also has a hinged lid, locking latch, and outside handles.
Specifications
  • Dimensions:
    8×14×11/4"