Cold Extractor, COEX, VELP Scientifica
COEX Cold Extractor, 230V/60 Hz
Catalog # 10184-180
Jump to:
Discontinued
Product Details & Documents
Cold extractor that performs rapid fat extraction directly in the same glass crucibles that are used by the FIWE Series for raw fiber analysis.
- Precise Results
- Versatile
- Convenient
Precise Results–Reduces fat to below 1% improving analytical precision. No loss – reduced sample remains in same crucible for analysis.
Versatile–Immediate raw fiber extraction after fat extraction here. Processing of individual or several samples.
Convenient–Fatty components collected for easy disposal. All reagents can be added through the same channel.
: Combine with FIWE for raw fiber analyzer.
Specifications
- Description:COEX Cold Extractor, 230V/60 Hz
- Cat. No.:10184-180
- W×H×D:730×300×380 mm
- Power:120W
- Weight:19 kg



