Order Entry
United States
Orders LinkContactUsLinkComponent
Dehydrant Blends, Epredia

Dehydrant Blends, Epredia

Dehydrant 95%, Poly Bottle, 1.3 gal.
Catalog # 10158-872
Supplier:   Epredia
Supplier #:  6315
Dehydrant Blends, Epredia
Catalog # 10158-872
Supplier:  Epredia
CAS Number:  
Dehydrant Blends, Epredia
Dehydrant Blends, Epredia

Dehydrant 95%, Poly Bottle, 1.3 gal.

CAS Number:  
Price
$235.03
New Customer Promotional Price
Log in for availability
Case of 4 (5l)
Product Details & Documents

Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.

  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents

Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.

Specifications

Specifications

  • Pack type:
    Poly Bottle
  • Description:
    Dehydrant 95%
  • Size:
    1.3 gal.

Specifications