Falcon® 96-Multiwell Insert Systems, Corning
Falcon™ 96-Multiwell Insert System with Feeder Tray and Lid, 96, 0.08 cm², 1.0 µm, Sterile, Tissue culture (TC)
Catalog # 10147-798
Supplier: Corning
Supplier #: 353938
Falcon® 96-Multiwell Insert Systems, Corning
Catalog # 10147-798
Supplier: Corning
Jump to:
Product Details & Documents
The Falcon® HTS 96-multiwell insert system is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays. The system has been tested for its ability to produce a differentiated monolayer of Caco-2, LLC-PK1, and MDCK cells making it an ideal platform for in vitro bioavailability and permeability studies. This automation compatible platform is composed of a 1.0 µm pore size PET membrane-based 96-multiwell insert plate, a media feeder tray and a lid. The drop-in baffle for the feeder tray mitigates media sloshing and lowers the risk of contamination.
- Automation compatible design suitable for use with most robots and fluid handling instruments
- Excellent reproducibility - the one-piece feeder tray enhances consistency in well-to-well monolayer growth
- Ideal for drug transport studies
Specifications
- No. of wells:96
- Pore size:1.0 µm
- Sterility:Sterile
- Description:Falcon™ 96-Multiwell Insert System with Feeder Tray and Lid
- Surface treatment:Tissue culture (TC)
- Cell growth area:0.08 cm²
- Cat. No.:10147-798
- Environmental Certification:ISO 14001 - Bedford (reg. no. 14.4942)
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #AvailabilityPrice
- Falcon™ 96 square well, angled bottom liquid receiver tray and lidFalcon™ 96 square well, angled bottom liquid receiver tray and lidBD-353925Log in for availabilityCase of 5 - $170.17



