Bluetooth Serial Adapter
Bluetooth dongle v2.0 RS232 single serial adapter ADP-BT-S for wireless connection
Catalog # 10147-670
Supplier: Mettler Toledo
Supplier #: 30086494
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
This paired bluetooth RS232 serial adapter ADP-BT-S is specially designed to provide wireless connection between printer and balance in laboratory weighing applications.
- Efficient use of workspace
This ensures error-free data transfer by simply plug in the BT adapter to wirelessly send data between your balance and a PC, printer or other peripheral device.
Specifications
- Description:Bluetooth dongle v2.0 RS232 single serial adapter ADP-BT-S for wireless connection