Silver Nitrate
Silver Nitrate, Solution, Laboratory, 100 mL
Supplier: Ward's Science
Supplier #: SS0162-100ML
Jump to:
Product Details & Documents
CAS Number: 7761-88-8
Formula: AgNO3
Density: 4.35 g/mL
Freezing Point: 212 °C
Solubility: Water, Hot Alcohol
Shelf Life: 24 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: This product is a corrosive, toxic irritant.
Documents
sds Silver Nitrate, 0.05 M Solution ,SS0183.pdf sds_Silver Nitrate, 0.0282 Molar (0.0282 Normal) Solution_SS0175.pdf sds_Silver Nitrate, 0.05 Molar (0.05 Normal) Solution_SS0183.pdf sds_Silver Nitrate, 0.1 Molar (0.1 Normal) Solution_SS0180.pdf sds_Silver Nitrate, 0.5 Molar (0.5 Normal) Solution_SS0181.pdf
SDS
More
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Description:Silver Nitrate, Solution
- Size:100 mL
- Chemical/physical specifications:1%
- Grade:Laboratory



