Order Entry
United States
Orders LinkContactUsLinkComponent
StabilZyme® SELECT Stabilizer, Surmodics

StabilZyme® SELECT Stabilizer, Surmodics

StabilZyme SELECT® Stabilizer, HRP/General, 50 mL
Catalog # 102466-328
Marketsource
Supplier:   Surmodics
Supplier #:  SZ03-0050
StabilZyme® SELECT Stabilizer, Surmodics
Catalog # 102466-328
Supplier:  Surmodics
Marketsource
CAS Number:  
StabilZyme® SELECT Stabilizer, Surmodics

StabilZyme SELECT® Stabilizer, HRP/General, 50 mL

CAS Number:  
Price
$40.00
Log in for availability
Each (50ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The StabilZyme® SELECT Stabilizer is a multi-purpose stabilizing solution designed to maintain the conformation of antibody/antigen conjugates, antibody coated particles and unmodified proteins that are commonly used in immunoassays. Adding the conjugate/protein directly into the SELECT Stabilizer allows for stable storage of the conjugate at a working concentration and eliminates the need for subsequent dilutions.

  • Long-term in solution stability
  • longer shelf life for kits
  • 2.5 year shelf life

StabilZyme® SELECT Stabilizer is a general purpose in-solution protein stabilizer that can be used for HRP conjugates, standards and controls in immunoassay applications.

Certifications: ISO 13485:2016 & 9001:2015 Certified

Specifications

Specifications

  • Description:
    StabilZyme SELECT® Stabilizer
  • Size:
    50 mL
  • Conjugate:
    HRP/General
  • Cat. No.:
    102466-328
  • Storage Buffer:
    None
  • pH:
    6.5-7.5
  • Shelf Life:
    2.5 years
  • Application:
    ELISA, Western Blot, ImmunoBlot
  • Protein:
    Bovine
  • Storage Temperature:
    2 to 8 °C or Room Temperature

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.