Order Entry
United States
ContactUsLinkComponent
RBC LYSIS BUFFER (10X) 100 ML
RBC Lysis Buffer (10X), Tonbo Biosciences
Catalog # 10141-140
Supplier:  Tonbo Biosciences
CAS Number:  
undefined
RBC Lysis Buffer (10X), Tonbo Biosciences
Catalog # 10141-140
Supplier:  Tonbo Biosciences
Supplier Number:  TNB-4300-L100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 mL
  • Storage Temperature:
    Store Undiluted at 2 to 8°C
  • Cat. No.:
    10141-140
  • Supplier no.:
    TNB-4300-L100
  • Reactivity:
    Mouse, Human

Specifications

About this item

RBC Lysis Buffer (10X) is a concentrated ammonium chloride-based lysing reagent

The diluted 1X working solution will lyse red blood cells in single cell suspensions with minimal effects on leukocytes. RBC Lysis Buffer (10X) does not contain a fixative so the cells remain viable after red blood cell lysis.

Frequently Bought Together