Spray Chambers, PerkinElmer
O-ring free chamber for oil analysis (4 mm baffle)
Catalog # 10138-186
Supplier: PerkinElmer
Supplier #: N0775355
Jump to:
Product Details & Documents
The spray chamber is an integral part of the sample introduction system responsible for filtering the sample mist to permit the appropriate droplet size distribution to reach the plasma.
The Scott spray chamber is corrosion-resistant and is inert to most mineral acids including HF, aqua regia and all organic solvents normally used in ICP analyses.
Specifications
- Description:O-ring free chamber for oil analysis (4 mm baffle)