Orders LinkContactUsLinkComponent
Spray Chambers, PerkinElmer
O-ring free chamber
  10138-182
 :  N0775351
Spray Chambers, PerkinElmer
  10138-182
Spray Chambers, PerkinElmer
Spray Chambers, PerkinElmer
O-ring free chamber
$2,192.42
Each
The spray chamber is an integral part of the sample introduction system responsible for filtering the sample mist to permit the appropriate droplet size distribution to reach the plasma.
The Scott spray chamber is corrosion-resistant and is inert to most mineral acids including HF, aqua regia and all organic solvents normally used in ICP analyses.
  • Description:
    O-ring free chamber