Order Entry
United States
Orders LinkContactUsLinkComponent
Chloroform, GR for analysis, Supelco®

Chloroform, GR for analysis, Supelco®

Glass bottle, 1 L
Catalog # EM1.02442.1000
Toxic
Health
Supplier:   MilliporeSigma
Supplier #:  1024421000
Chloroform, GR for analysis, Supelco®
Catalog # EM1.02442.1000
Supplier:  MilliporeSigma
CAS Number:  67-66-3
Chloroform, GR for analysis, Supelco®
Chloroform, GR for analysis, Supelco®

Glass bottle, 1 L

CAS Number:  67-66-3
Price
$140.82
Log in for availability
Each (1l) Case of 6 (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61.7 °C (1013 hPa)
  • Melting Pt:–63 °C
  • Density:1.4832 g/cm³ (20 °C)
  • EINECS:200-663-8
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • UN:1888
  • REACH:05-2114614906-45

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.