Order Entry
United States
Orders LinkContactUsLinkComponent
Fluid A, USP

Fluid A, USP

Fluid A, USP, 237 ml, 90 ml, Jar, 3
Catalog # 10135-872
Supplier #:  U17
Fluid A, USP
Catalog # 10135-872
Supplier:  
CAS Number:  
Fluid A, USP
Fluid A, USP

Fluid A, USP, 237 ml, 90 ml, Jar, 3

CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A rinsing and diluting fluid for use in USP testing protocols.

A rinsing and diluting fluid for use in USP testing protocols. Contains peptone 0.1%.

Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center. Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.

Ordering information: Order by the pack of 10

Specifications

Specifications

  • Pack type:
    Jar
  • Description:
    Fluid A, USP
  • Size:
    237 ml
  • Minimum Order Quantity:
    3
  • Volume:
    90 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.