Order Entry
United States
Orders LinkContactUsLinkComponent
Trypsin/ EDTA

Trypsin/ EDTA

Trypsin (0.25%), EDTA (0.02%), Membrane Filtered; prepared with Hanks' Balanced Salt Solution (HBSS) without calcium and magnesium, 100 mL
Catalog # 10128-794
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Supplier #:  118-093-721EA
Trypsin/ EDTA
Catalog # 10128-794
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Trypsin/ EDTA
Trypsin/ EDTA

Trypsin (0.25%), EDTA (0.02%), Membrane Filtered; prepared with Hanks' Balanced Salt Solution (HBSS) without calcium and magnesium, 100 mL

CAS Number:  
Price
$49.27
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Trypsin is a proteolytic enzyme used to detach adherent cell from culture vessel surfaces. Typical use includes removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.

  • With EDTA
  • With Phenol Red

Trypsin is a serine protease derived from porcinepancreas. It is a single chain polypeptide of 223 amino acid residue with substrate specificity based on positively charged Lysine and Arginine side chains. Trypsin predominantly cleaves peptide chains at the carboxyl sides of Lysine and Arginine, except when either is followed by Proline. It is most commonly used for dissociation and disaggregation of adherent cells.

Ethylenediaminetetraacetic acid (EDTA), a chelating agent often added to enhance enzymatic activity of trypsin solution. EDTA acts by neutralizing calcium and magnesium ions that enhance cell-to-cell adhesion. Polyvinylpyrrolidone (PVP) a synthetic polymer is known to increase cell viability especially in low serum or serum free condition.

Sterile Filtered
Performance Tested

Certifications: Manufactured in an ISO 9001:2015 certified facility.

Ordering information: *NOTE: During shipping, the trypsin might change color from pink to yellow due to exposure to carbon dioxide released from the dry ice in the shipping container. In our experience, this discoloration does not effect the functionality of the trypsin.

Caution: Research Use Only

Documents

Specifications

Specifications

  • Description:
    Trypsin (0.25%), EDTA (0.02%)
  • Size:
    100 mL
  • Formulation:
    Membrane Filtered; prepared with Hanks' Balanced Salt Solution (HBSS) without calcium and magnesium
  • Storage Conditions:
    –10 °C…–20 °C
  • Cat. No.:
    10128-794
  • Shipping Conditions:
    on dry ice

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.