Order Entry
United States
Orders LinkContactUsLinkComponent
 

Trypsin/ EDTA

Trypsin (0.25%), EDTA (0.02%), Sterile filtered, contains Phenol Red, 4x100 ml
  10128-792
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  118-093-721
Trypsin/ EDTA
  10128-792
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
Trypsin/ EDTA
Trypsin/ EDTA

Trypsin (0.25%), EDTA (0.02%), Sterile filtered, contains Phenol Red, 4x100 ml

 :  
 
$165.10
   
Pack of 4 (100ml)
 
   
 

Trypsin is a proteolytic enzyme used to detach adherent cell from culture vessel surfaces. Typical use includes removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.

  • With EDTA
  • With Phenol Red

Trypsin is a serine protease derived from porcinepancreas. It is a single chain polypeptide of 223 amino acid residue with substrate specificity based on positively charged Lysine and Arginine side chains. Trypsin predominantly cleaves peptide chains at the carboxyl sides of Lysine and Arginine, except when either is followed by Proline. It is most commonly used for dissociation and disaggregation of adherent cells.

Ethylenediaminetetraacetic acid (EDTA), a chelating agent often added to enhance enzymatic activity of trypsin solution. EDTA acts by neutralizing calcium and magnesium ions that enhance cell-to-cell adhesion. Polyvinylpyrrolidone (PVP) a synthetic polymer is known to increase cell viability especially in low serum or serum free condition.

Sterile Filtered
Performance Tested

 : Manufactured in an ISO 9001:2015 certified facility.

 : *NOTE: During shipping, the trypsin might change color from pink to yellow due to exposure to carbon dioxide released from the dry ice in the shipping container. In our experience, this discoloration does not effect the functionality of the trypsin.

 : Research Use Only

 

 

 

  • Description:
    Trypsin (0.25%), EDTA (0.02%)
  • Size:
    4x100 ml
  • Formulation:
    Sterile filtered, contains Phenol Red
  • Storage Conditions:
    –10 °C…–20 °C
  • Cat. No.:
    10128-792
  • Shipping Conditions:
    on dry ice

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.