Order Entry
United States
Orders LinkContactUsLinkComponent
 

Dehydrant Blends, Epredia

Dehydrant 70%, Poly Bottle, 1.3 gal.
  10128-124
 :   Epredia
 :  6515
Dehydrant Blends, Epredia
  10128-124
 :  Epredia
 :  
Dehydrant Blends, Epredia
Dehydrant Blends, Epredia

Dehydrant 70%, Poly Bottle, 1.3 gal.

 :  
 
$237.90
 
   
Case of 4 (20l)
 

Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.

  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents

Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.

 

 

  • Pack type:
    Poly Bottle
  • Description:
    Dehydrant 70%
  • Size:
    1.3 gal.