Order Entry
United States
Orders LinkContactUsLinkComponent
 

Buffer Solution

pH 8, Laboratory, 500 mL
Catalog # 470300-496
Supplier:   Ward's Science
 :  BB0408-500ML
Buffer Solution
Catalog # 470300-496
Supplier:  Ward's Science
CAS Number:  
Buffer Solution
Buffer Solution

pH 8, Laboratory, 500 mL

CAS Number:  
Price
$12.05
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Buffer solution has a shelf life of 24 months

  • Density: Approximately 1 g/mL
  • Boiling Point: 100°C
  • Freezing Point: 0
  • Shelf Life: 24 Months
  • Storage: Green

Its density is 1 g/mL.

Buffer solution has a boiling point of 100°C and a freezing point of 0°C.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    pH 8
  • Size:
    500 mL
  • Chemical/physical specifications:
    Clear
  • Grade:
    Laboratory

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.