Ammonium Hydroxide
Ammonium hydroxide, solution, Laboratory, 1 L
: AA0275-1L
CAS Number: 1336-21-6
Formula: NH4OH
Density: 0.900 g/mL
Boiling and Freezing Point: 36°C, -77°C
Synonyms: Ammonium Hydroxide Water Solution
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
: Corrosive, irritant and toxic.
- Description:Ammonium hydroxide, solution
- Size:1 L
- Chemical/physical specifications:1.0 M
- Grade:Laboratory



