Order Entry
United States
Orders LinkContactUsLinkComponent
 

Buffer Solution

pH 11, Laboratory, 500 mL
  470300-502
 :  BB0411-500ML
Buffer Solution
  470300-502
 :  
Buffer Solution
Buffer Solution

pH 11, Laboratory, 500 mL

 :  
 
$12.80
   
Each (500l)
 
   
 

Buffer solution has a shelf life of 24 months

  • Density: Approximately 1 g/mL
  • Boiling Point: 100°C
  • Freezing Point: 0
  • Shelf Life: 24 Months
  • Storage: Green

Its density is 1 g/mL.

Buffer solution has a boiling point of 100°C and a freezing point of 0°C.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

 

 

  • Description:
    pH 11
  • Size:
    500 mL
  • Chemical/physical specifications:
    Clear
  • Grade:
    Laboratory

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.