Order Entry
United States
Orders LinkContactUsLinkComponent
 

Silver Nitrate

Silver Nitrate, Reagent ACS, 100 g
Catalog # 470302-402
Supplier:   Ward's Science
 :  SS0160-100GR
Silver Nitrate
Catalog # 470302-402
Supplier:  Ward's Science
CAS Number:  
Silver Nitrate
Silver Nitrate

Silver Nitrate, Reagent ACS, 100 g

CAS Number:  
Price
$1,750.00
Log in for availability
Each (100g)
 
Product Details & Documents

CAS Number: 7761-88-8
Formula: AgNO3
Density: 4.35 g/mL
Freezing Point: 212 °C
Solubility: Water, Hot Alcohol
Shelf Life: 24 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

 : This product is a corrosive, toxic irritant.

Specifications

Specifications

  • Description:
    Silver Nitrate
  • Size:
    100 g
  • Chemical/physical specifications:
    Crystal
  • Grade:
    Reagent ACS

Specifications